Learn More
Dengue Virus Envelope Antibody (1) [DyLight 405] - Pan Specific, (Domain III), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-43767V
Product Code. 30281467
  • 507.00 € / 0.10mL
Description
Dengue Virus Envelope Monoclonal antibody specifically detects Dengue Virus Envelope in Virus samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dengue Virus Envelope
Monoclonal
DyLight 405
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Virus
Purified
ELISA, Western Blot
1
Western Blot, ELISA
dengue virus polyprotein
This antibody was obtained from a rabbit immunized with purified, recombinant Dengue Virus Envelope (type 1, US/Hawaii/1944) (RefSeq#: ACF49259.1; Val580-Lys680).
0.1 mL
Primary
Store at 4°C in the dark.