missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus NS1 protein Antibody (2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06436-100ul
This item is not returnable.
View return policy
Description
Dengue Virus NS1 protein Monoclonal antibody specifically detects Dengue Virus NS1 protein in Virus samples. It is validated for ELISA
Specifications
| Dengue Virus NS1 protein | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dengue Virus NS1 protein (Accession#: AAC59275.1; Asp1-Ala352). | |
| 100 μg | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| ELISA | |
| 2 | |
| ELISA 1:1000-1:2000 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction