missing translation for 'onlineSavingsMsg'
Learn More

Denhardt's Reagent, 50X Powder, Molecular Biology, Fisher BioReagents

Product Code. 10192853
Click to view available options
Quantity:
150 mg
Unit Size:
150mg
This item is not returnable. View return policy

Product Code. 10192853

Brand: Fisher BioReagents BP5205

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 26873-85-8

Amber Glass; 150mg

  • Used to make a 50X concentrate solution that can be diluted for use as a blocking agent in nucleic acid hybridization techniques
  • DNase, RNase, and protease free powder that is pre-mixed for your convenience to make 500mL of 50X solution
  • Eliminates the need to separately weigh ingredients - simply dissolve powder in pure water
  • The 50X concentrated solution can be diluted for use in hybridization techniques
  • Commonly used as a blocking buffer for Southern and Northern blot membranes
TRUSTED_SUSTAINABILITY

Specifications

Absorbance 0.6 to 0.8 AU at 278nm
Chemical Name or Material Denhardt's Reagent
Name Note 50X Powder
CAS 26873-85-8, 9003-39-8, 9048-46-8
Synonym Denhardt's Solution
Grade Molecular Biology
Concentration or Composition (by Analyte or Components) 50 mg each of bovine serum albumin (BSA), polyvinylpyrrolidone, and Ficoll™400
Packaging Amber Glass
Specific Gravity Report
pH 5 to 7
Quantity 150 mg
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • Denhardt`s Reagent 50X Powder 150 mg
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.