missing translation for 'onlineSavingsMsg'
Learn More

Density Standard, ASTM D 4052; ISO 12185 (Nominal value: 0.7933 g/mL, 15 °C), Paragon Scientific™

Product Code. 16010040
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16010040 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16010040 Supplier Paragon Scientific Supplier No. CRMDEKR
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Density Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Density according to ASTM D4052. Alternative test methods are IP 365 and ISO 12185.

  • Certified Reference Material for Density
  • Manufactured in accordance with ASTM / EN or IP test methods
  • Certified via Round Robin
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom

Specifications

UN Number UN 1223
Quantity 250 mL
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Certified Reference Material Density Standard, Jet Aviation Fuel (nominal value0.8025g/mL @ 15°C)
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
  • P391-Collect spillage.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: Kerosine sweetened

Nominal values quoted. Certified Values will vary from batch to batch.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.