missing translation for 'onlineSavingsMsg'
Learn More

Deoxy Epinephrine Hydrochloride, TRC

Product Code. 30008376
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008376 2.5 g 2.5g
30005429 250 mg 250mg
30004169 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008376 Supplier Toronto Research Chemicals Supplier No. TRCD2329202.5G
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-32-8
Molecular Formula C9 H13 N O2 . Cl H
Molecular Weight (g/mol) 203.67
Synonym 1,2-Benzenediol, 4-[2-(methylamino)ethyl]-, hydrochloride (1:1), 1,2-Benzenediol, 4-[2-(methylamino)ethyl]-, hydrochloride (9CI), Pyrocatechol, 4-[2-(methylamino)ethyl]-, hydrochloride (8CI), 3,4-Dihydroxyphenylethylmethylamine hydrochloride, 4-(2-Methylaminoethyl)-1,2-benzenediol hydrochloride, 4-(2-Methylaminoethyl)pyrocatechol hydrochloride, Deoxyepinephrine hydrochloride, Ephinine hydrochloride, Epinine hydrochloride, Methyl[β-(3,4-dihydroxyphenyl)ethyl]amine hydrochloride, Methyldopamine hydrochloride, N-Methyldopamine hydrochloride
IUPAC Name 4-[2-(methylamino)ethyl]benzene-1,2-diol;hydrochloride
SMILES Cl.CNCCc1ccc(O)c(O)c1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C9H13NO2.ClH/c1-10-5-4-7-2-3-8(11)9(12)6-7;/h2-3,6,10-12H,4-5H2,1H3;1H
Formula Weight 203.0713
Percent Purity >95%
Chemical Name or Material Deoxy Epinephrine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.