missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deoxyguanosine kinase Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92801-0.02ml
This item is not returnable.
View return policy
Description
Deoxyguanosine kinase Polyclonal antibody specifically detects Deoxyguanosine kinase in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Deoxyguanosine kinase | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| deoxyguanosine kinase, deoxyguanosine kinase, mitochondrial, DGK, dGKmitochondrial deoxyguanosine kinase, EC 2.7.1.113, MTDPS3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-277 of human DGUOK (NP_550438.1). RLSIEGNIAVGKSTFVKLLTKTYPEWHVATEPVATWQNIQAAGTQKACTAQSLGNLLDMMYREPARWSYTFQTFSFLSRLKVQLEPFPEKLLQARKPVQIFERSVYSDRYIFAKNLFENGSLSDIEWHIYQDWHSFLLWEFASRITLHGFIYLQASPQVCLKRLYQRAREEEKGIELAYLEQLHGQHEAWLIHKTTKLHFEALMNIPVLVLDVNDDFSEEVTKQEDLMREVNTFVKNL | |
| 0.02 mL | |
| Signal Transduction | |
| 1716 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction