missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Derlin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
244.00 € - 489.00 €
Specifications
| Antigen | Derlin 1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry-Paraffin 5-10 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18797003
|
Novus Biologicals
NB100-448SS |
0.025 mL |
244.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18737253
|
Novus Biologicals
NB100-448 |
0.1 mL |
489.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Derlin 1 Polyclonal specifically detects Derlin 1 in Human, Mouse, Canine, Chinese Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Derlin 1 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Polyclonal | |
| Rabbit | |
| ER Markers, Immunology, Lipid and Metabolism, Protein Turnover, Signal Transduction, Unfolded Protein Response | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| DER-1, DER1Degradation in endoplasmic reticulum protein 1, Der1-like domain family, member 1, Der1-like protein 1, derlin-1, DERtrin-1, FLJ13784, FLJ42092, MGC3067, PRO2577 | |
| DERL1 | |
| IgG | |
| Affinity Purified | |
| Human, Canine and Chinese Hamster. Predicted to react with Xenopus, bovine, chicken, rat, zebrafish, and mouse based on 100% sequence homology. Mouse reactivity reported in scientific literature (PMID: 25377857). |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Canine, Chinese Hamster | |
| Q9BUN8 | |
| 79139 | |
| A synthetic peptide made to the C-terminal region of human Derlin 1 (between residues 200-251). [UniProt# Q9BUN8] | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 29 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title