missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DERP6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85250
This item is not returnable.
View return policy
Description
DERP6 Polyclonal specifically detects DERP6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| DERP6 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| chromosome 17 open reading frame 81, dermal papilla derived protein 6, dermal papilla-derived protein 6, DERP6, MST071, MSTP071, S-phase 2 protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ELP5 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GTGRELEMLDSLLALGGLVLLRDSVEWEGRSLLKALVKKSALCGEQVHILGCEVSEEEFREGFDSDINNRL | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 23587 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction