missing translation for 'onlineSavingsMsg'
Learn More

Desacetyl Rifampicin Quinone (>85%), TRC

Product Code. 30133979
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133979 50 mg 50mg
30004615 25 mg 25mg
30005880 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133979 Supplier Toronto Research Chemicals Supplier No. TRCD19955550MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65110-92-1
Molecular Formula C41H54N4O11
Molecular Weight (g/mol) 778.89
Synonym 25-Deacetyl-1,4-didehydro-1,4-dideoxy-3-[[(4-methyl-1-piperazinyl)imino]methyl]-1,4-dioxo-Rifamycin, 25-Desacetylrifampin Quinone;
IUPAC Name (7S,9E,11S,12S,13S,14R,15R,16R,17S,18S,19E,21E)-2,13,15,17-tetrahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-26-[(Z)-(4-methylpiperazin-1-yl)iminomethyl]-8,30-dioxa-24-azatetracyclo[23.3.1.14,7.05,28]triaconta-1(28),2,4,9,19,21,25-heptaene-6,23,27,29-tetrone
SMILES C[C@H]1/C=C/C=C(/C(=O)N=C2/C(=C\NN3CCN(CC3)C)/C(=O)C4=C5C(=C(C(=C4C2=O)O)C)O[C@@](C5=O)(O/C=C/[C@@H]([C@H]([C@H]([C@@H]([C@@H]([C@@H]([C@H]1O)C)O)C)O)C)OC)C)\C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C41H54N4O11/c1-20-11-10-12-21(2)40(53)43-31-26(19-42-45-16-14-44(8)15-17-45)36(50)28-29(37(31)51)35(49)25(6)38-30(28)39(52)41(7,56-38)55-18-13-27(54-9)22(3)33(47)24(5)34(48)23(4)32(20)46/h10-13,18-20,22-24,27,32-34,42,46-49H,14-17H2,1-9H3/b11-10+,18-13+,21-12+,26-19+,43-31?/t20-,22+,23+,24-,27-,32-,33+,34+,41-/m0/s1
Formula Weight 778.3789
Chemical Name or Material Desacetyl Rifampicin Quinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.