missing translation for 'onlineSavingsMsg'
Learn More

Desaminotyrosine, MedChemExpress

Product Code. 16459376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459376 Supplier Medchem Express Supplier No. HYW015346
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Desaminotyrosine is a microbially associated metabolite protecting from influenza through augmentation of type I interferon signaling.

Desaminotyrosine is a microbially associated metabolite protecting from influenza through augmentation of type I interferon signaling.

Specifications

Chemical Name or Material Desaminotyrosine
CAS 501-97-3
Color White
Purity Grade Notes Research
Molecular Formula C9H10O3
Quantity 100 mg
Synonym 3-(4-Hydroxyphenyl)propionic acid
Solubility Information DMSO : 100 mg/mL (601.76 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (200.57 mM; Need ultrasonic)
SMILES C1=C(C=CC(=C1)O)CCC(O)=O
Molecular Weight (g/mol) 166.18
Formula Weight 166.18
Percent Purity 98.46%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.