missing translation for 'onlineSavingsMsg'
Learn More

Desidustat, MedChemExpress

Product Code. 16494856
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494856 100 mg 100mg
16483808 10 mg 10mg
16460187 50 mg 50mg
16446537 5 mg 5mg
16400999 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494856 Supplier Medchem Express Supplier No. HY103227

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Desidustat is an inhibitor of HIF hydroxylase extracted from patent WO 2014102818 A1, compound example 2.

Desidustat is an inhibitor of HIF hydroxylase extracted from patent WO 2014102818 A1, compound example 2.

Specifications

Chemical Name or Material Desidustat
Physical Form Solid
CAS 1616690-16-4
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C16H16N2O6
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 10 mg/mL (30.09 mM; Need ultrasonic)
SMILES O=C(O)CNC(C1=C(O)C2=C(N(OCC3CC3)C1=O)C=CC=C2)=O
Molecular Weight (g/mol) 332.31
Formula Weight 332.31
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.