missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmethyl Erlotinib hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Desmethyl Erlotinib hydrochloride (OSI-420) is an active metabolite of Erlotinib. Erlotinib is a potent EGFR tyrosin kinase inhibitor.
Specifications
Specifications
| Chemical Name or Material | Desmethyl Erlotinib hydrochloride |
| CAS | 183320-51-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H22ClN3O4 |
| Quantity | 50 mg |
| Synonym | OSI-420CP-373420 hydrochloride |
| Solubility Information | DMSO : 50 mg/mL (120.23 mM; Need ultrasonic) |
| SMILES | OCCOC1=CC2=C(N=CN=C2NC3=CC=CC(C#C)=C3)C=C1OCCOC.Cl |
| Molecular Weight (g/mol) | 415.87 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction