missing translation for 'onlineSavingsMsg'
Learn More

Desmethylnortriptyline, TRC

Product Code. 30350416
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30350416 5 mg 5mg
30350056 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30350416 Supplier Toronto Research Chemicals Supplier No. TRCD2930755MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4444-42-2
Molecular Formula C18H19N
Molecular Weight (g/mol) 249.35
Synonym 3-(10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-1-propanamine, 10,11-Dihydro-5H-dibenzo[a,d]cycloheptene-Δ5,γ-propylamine, Demethylnortriptyline, Didesmethylamitriptyline, Lu 5-079, N,N-Didemethylamitriptyline
SMILES NCC/C=C1C2=CC=CC=C2CCC3=C/1C=CC=C3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H19N/c19-13-5-10-18-16-8-3-1-6-14(16)11-12-15-7-2-4-9-17(15)18/h1-4,6-10H,5,11-13,19H2
Chemical Name or Material Desmethylnortriptyline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.