missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmocollin-1 Antibody (SR1004), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22640-50ul
This item is not returnable.
View return policy
Description
Desmocollin-1 Monoclonal antibody specifically detects Desmocollin-1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Desmocollin-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1823 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR1004 | |
| Western Blot 1:500-1:2000 | |
| cadherin family member 1, CDHF1, desmocollin 1, desmocollin-1, desmosomal glycoprotein 2/3, DG2/DG3 | |
| A synthesized peptide derived from human Desmocollin-1 (Uniprot #: Q08554) | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction