missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Desmoglein 4 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149224
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Desmoglein proteins are cadherin-type cell adhesion molecules. Desmoglein 4 (dsg4) shares 41% identity with human desmoglein 1, 37% with human desmoglein 2 and 50% with human desmoglein 3. A type I membrane protein of the cadherin protein family, dsg4 is expressed in salivary gland, suprabasal epidermis, hair follicle, testis, prostate and skin. In the hair follicle, dsg4 is an important mediator of keratinocyte cell adhesion and coordinates the transition from proliferation to differentiation. The human DSG4 gene is composed of 16 exons spanning approximately 37 kb of 18q12 and is situated between DSG1 and DSG3.
Specifications
| Desmoglein 4 | |
| Polyclonal | |
| Unconjugated | |
| DSG4 | |
| Cadherin family member 13; CDGF13; CDHF13; desmoglein 4; Desmoglein4; desmoglein-4; DSG4; HYPT6; LAH; lanceolate hair | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 147409 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q86SJ6 | |
| DSG4 | |
| Mouse myeloma cell line NS0-derived recombinant human Desmoglein-4, Glu24-Ala631. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction