missing translation for 'onlineSavingsMsg'
Learn More

Dess-Martin periodinane, 15 wt.% solution in dichloromethane

Product Code. 15284704 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15284704 250 mL 250mL
10562861 50 mL 50mL
15394978 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15284704 Supplier Thermo Scientific Acros Supplier No. 333112500
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

Dess-Martin periodinane, 10 to 15%, C13H13IO8, CAS Number-87413-09-0, 75-09-2, dess martin, dess-martin, dess-martinperiodinane, dess-martin periodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess martin periodinane, dess-martin periodane, dess-martin reagent, triacetoxyperiodinane | CAS: 87413-09-0 | C13H13IO8 | 424.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

oxidation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dess-Martin periodinane
Color Colorless to Yellow
Name Note 15 wt. % Solution in Dichloromethane
CAS 87413-09-0, 75-09-2
Molecular Formula C13H13IO8
MDL Number MFCD00130127
Density 1.3620g/mL
Synonym dess-martin periodinane, triacetoxyperiodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess-martin reagent, dess-martinperiodinane, dess-martin, dess martin periodinane, dess martin, dess-martin periodane
InChI Key NKLCNNUWBJBICK-UHFFFAOYSA-N
SMILES CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12
IUPAC Name (1,1-diacetyloxy-3-oxo-1$l^{5},2-benziodoxol-1-yl) acetate
Molecular Weight (g/mol) 424.14
PubChem CID 159087
Formula Weight 424.15
Percent Purity 10 to 15%
Specific Gravity 1.362
Quantity 250 mL
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.