missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Destrin Monoclonal Antibody (24GB5625)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557389
This item is not returnable.
View return policy
Description
Destrin Monoclonal Antibody for Western Blot, Flow
The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| Destrin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P60981 | |
| DSTN | |
| Recombinant protein of human DSTN (aa 1-165). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5625 | |
| Unconjugated | |
| DSTN | |
| 2610043P17Rik; ACTDP; actin depolymerizing factor; actin-depolymerizing factor; ADF; AU042046; bA462D18.2; bA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)); corn1; corneal disease 1; destrin; destrin (actin depolymerizing factor); destrin, actin depolymerizing factor; Dsn; DSTN; epididymis luminal protein 32; HEL32; RP23-77A2.1; sID; Sid 23; Sid23; sid23p | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11034 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction