Learn More
Invitrogen™ Destrin Monoclonal Antibody (24GB5625)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA557390
 View more versions of this product
Product Code. 30276063
  • 198.00 € / 20µL
Description
Destrin Monoclonal Antibody for Western Blot, Flow
The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Destrin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P60981
DSTN
Recombinant protein of human DSTN (aa 1-165).
20 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Western Blot, Immunocytochemistry
24GB5625
Unconjugated
DSTN
2610043P17Rik; ACTDP; actin depolymerizing factor; actin-depolymerizing factor; ADF; AU042046; bA462D18.2; bA462D18.2 (destrin (actin depolymerizing factor ADF) (ACTDP)); corn1; corneal disease 1; destrin; destrin (actin depolymerizing factor); destrin, actin depolymerizing factor; Dsn; DSTN; epididymis luminal protein 32; HEL32; RP23-77A2.1; sID; Sid 23; Sid23; sid23p
Mouse
Affinity chromatography
RUO
11034
-20°C