missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexras1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92235-0.02ml
This item is not returnable.
View return policy
Description
Dexras1 Polyclonal antibody specifically detects Dexras1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Dexras1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| Activator of G-protein signaling 1, AGS1ras-related protein, DEXRAS1dexamethasone-induced Ras-related protein 1, MGC:26290, RAS, dexamethasone-induced 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 132-281 of human RASD1 (NP_057168.1). TKENVDVPLVICGNKGDRDFYREVDQREIEQLVGDDPQRCAYFEISAKKNSSLDQMFRALFAMAKLPSEMSPDLHRKVSVQYCDVLHKKALRNKKLLRAGSGGGGGDPGDAFGIVAPFARRPSVHSDLMYIREKASAGSQAKDKERCVIS | |
| 0.02 mL | |
| Signal Transduction | |
| 51655 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction