missing translation for 'onlineSavingsMsg'
Learn More

Dextran sulfate sodium salt, MW ca 40,000, Thermo Scientific™

Dextran sulfate sodium salt (benzocaine or sodium dextran sulfate, CAS # 94-09-7) is a glucose polymer of bacterial origin containing 17-20% sulfur.

Brand:  Thermo Scientific Alfa Aesar J63606.14

Quantity 25 g

 View more versions of this product

Product Code. 15435799

  • 78.30 € / 25g

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

  • Dextran sulfate sodium salt is the ethyl ester of p-aminobenzoic acid
  • Dextran sulfate inhibits voltage-dependent sodium channels, preventing the transduction of pain and itch
  • Dextran sulfate is speculated to have antiviral and antioxidant properties

Application

  • Dextran sulfate is suitable for biochemistry, cell culture, and in vivo work
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

9011-18-1
165.192
BLFLLBZGZJTVJG-UHFFFAOYSA-N
2337
ethyl 4-aminobenzoate
C9H11NO2
MFCD00081551
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
CHEBI:116735
CCOC(=O)C1=CC=C(C=C1)N

Specifications

9011-18-1
MFCD00081551
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
25 g
CCOC(=O)C1=CC=C(C=C1)N
165.192
CHEBI:116735
Phenol-like
C9H11NO2
142951
Soluble in water.
BLFLLBZGZJTVJG-UHFFFAOYSA-N
ethyl 4-aminobenzoate
2337
≈40 kd
Dextran sulfate sodium salt
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.