missing translation for 'onlineSavingsMsg'
Learn More

Dextran sulfate sodium salt, MW ca 40,000, Thermo Scientific™

Dextran sulfate sodium salt (benzocaine or sodium dextran sulfate, CAS # 94-09-7) is a glucose polymer of bacterial origin containing 17-20% sulfur.

78.30 € - 239.00 €

Chemical Identifiers

CAS 9011-18-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name ethyl 4-aminobenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15435799
SDS View Documents
Thermo Scientific Alfa Aesar
J63606.14
25 g
78.30 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15445799
SDS View Documents
Thermo Scientific Alfa Aesar
J63606.22
100 g
239.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

  • Dextran sulfate sodium salt is the ethyl ester of p-aminobenzoic acid
  • Dextran sulfate inhibits voltage-dependent sodium channels, preventing the transduction of pain and itch
  • Dextran sulfate is speculated to have antiviral and antioxidant properties

Application

  • Dextran sulfate is suitable for biochemistry, cell culture, and in vivo work
Specifications

Chemical Identifiers

9011-18-1
165.192
BLFLLBZGZJTVJG-UHFFFAOYSA-N
2337
ethyl 4-aminobenzoate
C9H11NO2
MFCD00081551
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
CHEBI:116735
CCOC(=O)C1=CC=C(C=C1)N

Specifications

9011-18-1
MFCD00081551
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
BLFLLBZGZJTVJG-UHFFFAOYSA-N
ethyl 4-aminobenzoate
2337
≈40 kd
Dextran sulfate sodium salt
C9H11NO2
142951
Soluble in water.
CCOC(=O)C1=CC=C(C=C1)N
165.192
CHEBI:116735
Phenol-like
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : HH9290000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.