missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DFNA5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSD00001W
This item is not returnable.
View return policy
Beschreibung
Glycerol (1:1) may be added for additional stability. Reconstitute in 100 μl sterile water. Centrifuge to remove any insoluble material.
Expressed in cochlea. Low level of expression in heart, brain, placenta, lung, liver, skeletal muscle, kidney and pancreas, with highest expression in placenta. Defects in DFNA5 are the cause of non-syndromic sensorineural deafness autosomal domit type
Spezifikation
| DFNA5 | |
| Polyclonal | |
| Gsdme | |
| Non-syndromic hearing impairment protein 5 homolog | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Q9Z2D3 | |
| Gsdme | |
| A synthetic peptide from mouse Dfna5 deafness | |
| RUO | |
| 54722 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur