missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGAT2 Antibody - BSA Free, Novus Biologicals™
Description
DGAT2 Polyclonal antibody specifically detects DGAT2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)
Specifications
Specifications
| Antigen | DGAT2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 - 2 μg/ml, Flow Cytometry 2.5 μg/ml, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 2 - 5 μg/ml, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) 2.5 μg/ml |
| Formulation | PBS |
| Gene Alias | diacylglycerol O-acyltransferase 2, diacylglycerol O-acyltransferase homolog 2, diacylglycerol O-acyltransferase homolog 2 (mouse), diacylglycerol O-acyltransferase-like protein 2, Diglyceride acyltransferase 2, DKFZp686A15125, EC 2.3.1, EC 2.3.1.20, GS1999FULL |
| Host Species | Rabbit |
| Immunogen | Partial recombinant protein made to an internal portion of the human DGAT2 protein (amino acids 268-377) [UniProt Q96PD7] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?