missing translation for 'onlineSavingsMsg'
Learn More

DGCR8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

450.00 € - 708.00 €

Specifications

Antigen DGCR8
Dilution Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18649594
View Documents
Novus Biologicals
NBP3-21407-25ul
25 μg
450.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18631095
View Documents
Novus Biologicals
NBP3-21407-100ul
100 μg
708.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DGCR8 Polyclonal antibody specifically detects DGCR8 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

DGCR8
Immunofluorescence
Unconjugated
Rabbit
Epigenetics, Stem Cell Markers
PBS, pH 7.2, 40% glycerol
54487
IgG
Affinity purified
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Polyclonal
Purified
RUO
Human
C22orf12, DGCRK6chromosome 22 open reading frame 12, DiGeorge syndrome critical region 8, DiGeorge syndrome critical region gene 8, Gy1, microprocessor complex subunit DGCR8, pasha
This antibody was developed against Recombinant Protein corresponding to amino acids: IDGVTYGSGTASSKKLAKNKAARATLEILIPDFVKQTSEEKPKDSEELEYFNHISIEDSRVYELTSKAGLLSPYQILHECLKRNHGMGDTSIKFEVVPG
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.