missing translation for 'onlineSavingsMsg'
Learn More

DHRSX Rabbit anti-Human, Polyclonal, R&D Systems™

Product Code. 18366522
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18366522 25 μg 25µL
18379165 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18366522 Supplier Bio-Techne Supplier No. NBP31783025UL
  • ONLINE EXCLUSIVE PRICE
    538.16 €
    458.85 € / 25µL
    Save 79.31 €
    14% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

DHRSX Polyclonal specifically detects DHRSX in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRSX
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias CXorf11, dehydrogenase/reductase (SDR family) X chromosome, dehydrogenase/reductase (SDR family) X-linked, DHRS5Xmember 1, EC 1.1, SDR46C1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: LDTLKESGSPGHSARVVTVSSATHYVAELNMDDLQSSACYSPHAAYAQSKLAL
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 207063
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.