missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHTKD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | DHTKD1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 4-8 uL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637125
|
Novus Biologicals
NBP2-99197-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605695
|
Novus Biologicals
NBP2-99197-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DHTKD1 Polyclonal antibody specifically detects DHTKD1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| DHTKD1 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 55526 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 4-8 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| dehydrogenase E1 and transketolase domain containing 1, Dehydrogenase E1 and transketolase domain-containing protein 1, DKFZp762M115, EC 1.2.4.2, KIAA1630DKFZP762M115, MGC3090, probable 2-oxoglutarate dehydrogenase E1 component DHKTD1, mitochondrial | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human DHTKD1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title