missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX36 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 463.00 €
Specifications
| Antigen | DHX36 |
|---|---|
| Dilution | Western Blot 1:200-1:2000, Immunoprecipitation 1:50-1:100 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641792
|
Novus Biologicals
NBP2-92933-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606712
|
Novus Biologicals
NBP2-92933-0.1ml |
0.1 mL |
463.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DHX36 Polyclonal antibody specifically detects DHX36 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| DHX36 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| DDX36, DEAH (Asp-Glu-Ala-His) box polypeptide 36, DEAH box protein 36, EC 3.6.1, EC 3.6.4.13, G4 resolvase-1, KIAA1488G4R1, MLEL1RHAUDEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 36, MLE-like protein 1, probable ATP-dependent RNA helicase DHX36, RNA helicase associated with AU-rich element ARE | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human DHX36 (NP_065916.2). MSYDYHQNWGRDGGPRSSGGGYGGGPAGGHGGNRGSGGGGGGGGGGRGGRGRHPGHLKGREIGMWYAKKQGQKNKEAERQERAVVHMDERREEQIVQLLNSVQAKNDKESEAQISWFAPEDHGYGTEVSTKNTPCSENKLDIQEKKLINQEKKMFRIRNRSYIDRDSEYLLQENEPDGTLDQKLLEDLQKKKNDLRYIEMQHFREKLPSYGMQKELVNLIDNHQVTVISGETGCGKTTQVTQFILDNYIERGKGSACRIV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:200-1:2000, Immunoprecipitation 1:50-1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 170506 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title