Learn More
Invitrogen™ DHX36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592353
Description
Immunogen sequence: MSYDYHQNWG RDGGPRSSGG GYGGGPAGGH GGNRGSGGGG GGGGGGRGGR GRHPGHLKGR EIGMWYAKKQ GQKNKEAERQ ERAVVHMDER REEQIVQLLN SVQAKNDKES EAQISWFAPE DHGYGTEVST KNTPCSENKL DIQEKKLINQ EKKMFRIRNR SYIDRDSEYL LQENEPDGTL DQKLLEDLQK KKNDLRYIEM QHFREKLPSY GMQKELVNLI DNHQVTVISG ETGCGKTTQV TQFILDNYIE RGKGSACRIV; Positive Samples: Mouse brain, Mouse liver, Mouse kidney, Rat brain; Cellular Location: Chromosome, Cytoplasm, Nucleus, telomere.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases implicated in several cellular processes involving modifications of RNA secondary structure. DHX36 (DEAH box protein 36), also known as MLE-like protein 1 and RNA helicase associated with AU-rich element ARE (RHAU), belongs to RNA helicase of the DEAH family and may function in sex development and spermatogenesis. It is expressed in testis and is evolutionary conserved with true orthologs in almost all animal species. DHX36 plays a role in degradation and deadenylation of mRNAs containing in their 3'-UTR the consensus ARE sequence element. DHX36 is required for early embryogenesis.
Specifications
| DHX36 | |
| Polyclonal | |
| Unconjugated | |
| DHX36 | |
| 2810407E23Rik; AI452301; Asp-Glu-Ala-His; ATP-dependent DNA/RNA helicase DHX36; ATP-dependent RNA helicase DHX36; AU022184; Ddx36; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 36; DEAD/H box polypeptide 36; DEAH (Asp-Glu-Ala-His) box polypeptide 36; DEAH box protein 36; DEAH-box helicase 36; DEAH-box protein 36; DHX36; G4 resolvase-1; G4R1; G4-resolvase 1; G4-resolvase-1; KIAA1488; mKIAA1488; Mlel1; MLE-like protein 1; probable ATP-dependent RNA helicase DHX36; RHAU; RNA helicase associated with AU-rich element ARE; RNA helicase associated with AU-rich element protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170506, 310461, 72162 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8VHK9, Q9H2U1 | |
| DHX36 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human DHX36 (NP_0659162). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.