Learn More
Invitrogen™ DHX9 Recombinant Rabbit Monoclonal Antibody (24GB4140)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557013
 View more versions of this product
Product Code. 30276200
  • 500.00 € / 100µL
Description
DHX9 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The human alanyl-tRNA synthetase (AARS) belongs to a family of tRNA synthases, of the class II enzymes. Class II tRNA synthases evolved early in evolution and are highly conserved. This is reflected by the fact that 498 of the 968-residue polypeptide human AARS shares 41% identity witht the E.coli protein. tRNA synthases are the enzymes that interpret the RNA code and attach specific aminoacids to the tRNAs that contain the cognate trinucleotide anticodons. They consist of a catalytic domain which interacts with the amino acid acceptor-T psi C helix of the tRNA, and a second domain which interacts with the rest of the tRNA structure.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DHX9
Recombinant Monoclonal
Unconjugated
Dhx9
AI326842; ATP-dependent RNA helicase A; Ddx9; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9; DEAH (Asp-Glu-Ala-His) box helicase 9; DEAH (Asp-Glu-Ala-His) box polypeptide 9; DEAH box protein 9; DEAH-box helicase 9; DExH-box helicase 9; Dhx9; FLJ17406; HEL-5; Leukophysin; LKP; mHEL-5; NDH II; NDH2; NDHII; Nuclear DNA helicase II; OTTHUMP00000033287; RHA; RNA helicase; RNA helicase A
Rabbit
Affinity chromatography
RUO
13211, 1660, 304859
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4140
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O70133, Q08211
Dhx9
A synthesized peptide derived from human RNA Helicase A (aa 1150-1200).
100 μL
Primary
Human, Mouse, Rat