missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butylamine, for HPLC
GREENER_CHOICE

Product Code. 10423745 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10423745 100 mL Glass bottle 100mL
10055343 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10423745 Supplier Thermo Scientific Supplier No. 390611000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-92-2 | C8H19N | 129.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-92-2
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.24
MDL Number MFCD00009429
InChI Key JQVDAXLFBXTEQA-UHFFFAOYSA-N
Synonym dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
PubChem CID 8148
IUPAC Name N-butylbutan-1-amine
SMILES CCCCNCCCC

Specifications

Melting Point -62.0°C
Color White
Density 0.7600g/mL
Boiling Point 159.0°C
Flash Point 39°C
Packaging Glass bottle
Linear Formula [CH3(CH2)3]2NH
Refractive Index 1.4160 to 1.4180
Quantity 100 mL
Residue after Evaporation 0.0005% max.
Merck Index 15, 3037
Specific Gravity 0.76
Solubility Information Solubility in water: 4.05g/L (25°C). Other solubilities: soluble in alcohol, ether, benzene, acetone, miscible with hydrocarbons
Transmittance 99 (c=0.01M water, 254nm) min.
Viscosity 0.9 mPa.s (20°C)
Formula Weight 129.24
Percent Purity ≥99% (GC)
Grade HPLC
Physical Form Crystalline Free Flowing Powder
Chemical Name or Material Di-n-butylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Di-n-butylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Di-n-butylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.