missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butylmagnesium, 0.5M solution in heptane, AcroSeal™

Product Code. 10609794 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10609794 800 mL AcroSeal™ Glass Bottle 800mL
10535793 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10609794 Supplier Thermo Scientific Acros Supplier No. 377778000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1191-47-5 | C8H18Mg | 138.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-47-5 , 142-82-5 , 97-93-8
Molecular Formula C8H18Mg
Molecular Weight (g/mol) 138.54
MDL Number MFCD00015225
InChI Key KJJBSBKRXUVBMX-UHFFFAOYSA-N
Synonym dibutylmagnesium, magnesium, dibutyl, di-n-butylmagnesium, magnesium,butyl 1-methylpropyl, dibutyl magnesium, di-n-butyl magnesium, di-n-butylmagnesium solution, odhfjiddbsdwnu-uhfffaoysa-n, di-n-butylmagnesium, 0,5m in heptane, di-n-butylmagnesium solution, 1.0 m in heptane
PubChem CID 70929
IUPAC Name magnesium;butane
SMILES [Mg++].CCC[CH2-].CCC[CH2-]

Specifications

Melting Point -71.0°C
Density 0.7300g/mL
Flash Point −4°C
Packaging AcroSeal™ Glass Bottle
Linear Formula (C4H9)2Mg
Quantity 800 mL
Specific Gravity 0.73
Solubility Information Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
Viscosity 0.9 mPa.s (22°C)
Formula Weight 138.54
Chemical Name or Material Di-n-butylmagnesium
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.