missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl phthalate, 98%

Product Code. 11430401
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430401 500 mL 500mL
11354268 25 mL 25mL
11364268 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430401 Supplier Thermo Scientific Alfa Aesar Supplier No. 041522.AP
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Plasticizer | CAS: 117-84-0 | C24H38O4 | 390.564 g/mol

Di-n-octyl phthalate is used as a plasticizer for many resins and elastomers. It acts as an additive. It is also used for medical tubing and blood storage bags, wire and cables, carpet back coating, floor tile and in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-84-0
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.564
MDL Number MFCD00015292
InChI Key MQIUGAXCHLFZKX-UHFFFAOYSA-N
Synonym dioctyl phthalate, di-n-octyl phthalate, dinopol nop, n-octyl phthalate, vinicizer 85, phthalic acid, dioctyl ester, polycizer 162, dnop, phthalic acid di-n-octyl ester, dioctyl 1,2-benzenedicarboxylate
PubChem CID 8346
ChEBI CHEBI:34679
IUPAC Name dioctyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCC

Specifications

Melting Point -25°C
Color Colorless to Yellow
Density 0.98 g/mL
Boiling Point 380°C
Flash Point 219°C (426°F)
Assay Percent Range 98%
Quantity 500 mL
Merck Index 14,2864
Solubility Information Insoluble in water.
Formula Weight 390.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Di-n-octyl phthalate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.