missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl Phthalate, TRC

Product Code. 30007171
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007171 10 g 10g
30004567 50 g 50g
17914995 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007171 Supplier Toronto Research Chemicals Supplier No. TRCD48175010G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-84-0
Molecular Formula C24 H38 O4
Molecular Weight (g/mol) 390.56
Synonym 1,2-Benzenedicarboxylic acid, 1,2-dioctyl ester, 1,2-Benzenedicarboxylic acid, dioctyl ester (9CI), Phthalic acid, dioctyl ester (6CI,8CI), 1,2-Di-n-octyl phthalate, Bis(n-octyl) phthalate, DNOP, Di-n-octyl phthalate, Dicaprylyl phthalate, Dinopol NOP, Dioctyl o-phthalate, Dioctyl phthalate, Monocizer W 555, NSC 15318, Octyl phthalate, Phthalic acid dioctyl ester, Vinycizer 85, n-Octyl phthalate, Phthalic acid di-n-octyl ester, Phthalic acid, bis-n-octyl ester
IUPAC Name dioctyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C24H38O4/c1-3-5-7-9-11-15-19-27-23(25)21-17-13-14-18-22(21)24(26)28-20-16-12-10-8-6-4-2/h13-14,17-18H,3-12,15-16,19-20H2,1-2H3
Formula Weight 390.277
Chemical Name or Material Di-n-octyl phthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.