missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propyl Phthalate, TRC

Product Code. 30012297
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012297 10 g 10g
30009474 50 g 50g
17970545 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012297 Supplier Toronto Research Chemicals Supplier No. TRCD49242510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-16-8
Formula Weight 250.1205
InChI Formula InChI=1S/C14H18O4/c1-3-9-17-13(15)11-7-5-6-8-12(11)14(16)18-10-4-2/h5-8H,3-4,9-10H2,1-2H3
IUPAC Name dipropyl benzene-1,2-dicarboxylate
Molecular Formula C14 H18 O4
Molecular Weight (g/mol) 250.29
Percent Purity >95%
SMILES CCCOC(=O)c1ccccc1C(=O)OCCC
Synonym 1,2-Benzenedicarboxylic acid, 1,2-dipropyl ester, 1,2-Benzenedicarboxylic acid, dipropyl ester (9CI), Phthalic acid, dipropyl ester (6CI,7CI,8CI), Di-n-Propyl phthalate, Dipropyl benzene-1,2-dicarboxylate, Dipropyl phthalate, NSC 15314, Phthalic acid dipropyl ester, Phthalic acid, bis-propyl ester
Chemical Name or Material Phthalic acid, bis-propyl ester
Grade HPLC
Show More Show Less

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.