missing translation for 'onlineSavingsMsg'
Learn More

Diacerein, MedChemExpress

Product Code. 16434807
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16434807 50 mg 50mg
16419046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16434807 Supplier Medchem Express Supplier No. HYN0283
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Diacerein (Diacerhein), a interleukin-1 beta inhibitor, is a slow-acting medicine of the class anthraquinone used to treat joint diseases.

Diacerein (Diacerhein), a interleukin-1 beta inhibitor, is a slow-acting medicine of the class anthraquinone used to treat joint diseases.

Specifications

Chemical Name or Material Diacerein
CAS 13739-02-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H12O8
Quantity 50 mg
Synonym Diacerhein Diacetylrhein
Solubility Information DMSO : 12.5 mg/mL (33.94 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C(C=C1C2=O)=CC(OC(C)=O)=C1C(C3=C2C=CC=C3OC(C)=O)=O)O
Molecular Weight (g/mol) 368.29
Formula Weight 368.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.