missing translation for 'onlineSavingsMsg'
Learn More

Diallyldibutyltin, Thermo Scientific™

Product Code. 11334567
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334567 2 g 2g
11354567 0.5 g 0.5g
11344567 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334567 Supplier Thermo Scientific Alfa Aesar Supplier No. 039621.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15336-98-8
Molecular Formula C14H28Sn
Molecular Weight (g/mol) 315.088
MDL Number MFCD00014944
InChI Key LCGVYMRFNOWPGQ-UHFFFAOYSA-N
Synonym diallyldibutyltin, diallyldibutylstannane, dibutylbis prop-2-en-1-yl stannane, diallyl dibutyl stannane, diallyldi-n-butyltin, acmc-1bvvj, dibutyl-bis prop-2-enyl stannane
PubChem CID 84873
IUPAC Name dibutyl-bis(prop-2-enyl)stannane
SMILES CCCC[Sn](CCCC)(CC=C)CC=C

Specifications

Density 1.1002 g/mL
Boiling Point 81°C (0.2 mmHg)
Refractive Index 1.5
Quantity 2 g
UN Number UN2788
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 315.07
Physical Form Liquid
Chemical Name or Material Diallyldibutyltin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.