missing translation for 'onlineSavingsMsg'
Learn More

Diatrizoic acid

Product Code. 15435689
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435689 25 g 25g
15425689 5 g 5g
15445689 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15435689 Supplier Thermo Scientific Alfa Aesar Supplier No. J63444.14
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

A high-osmolality radiocontrast agent | CAS: 117-96-4 | C11H9I3N2O4 | 613.92 g/mol

Diatrizoic acid is a gradient reagent used for blood cell separation and diagnostic aid (radiopaque medium). Diatrizoic acid may be used as an alternative to barium sulfate for medical imaging of the gastrointestinal tract, such as upper gastrointestinal series and small bowel series.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diatrizoic acid is a gradient reagent used for blood cell separation and diagnostic aid (radiopaque medium). Diatrizoic acid may be used as an alternative to barium sulfate for medical imaging of the gastrointestinal tract, such as upper gastrointestinal series and small bowel series.

Solubility
Soluble in Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-96-4
Molecular Formula C11H9I3N2O4
Molecular Weight (g/mol) 613.92
MDL Number MFCD00069960
InChI Key YVPYQUNUQOZFHG-UHFFFAOYSA-N
Synonym diatrizoic acid, amidotrizoic acid, diatrizoate, amidotrizoate, urogranoic acid, urografin acid, odiston, urotrast, diat, diatriazoate
PubChem CID 2140
ChEBI CHEBI:53691
IUPAC Name 3,5-diacetamido-2,4,6-triiodobenzoic acid
SMILES CC(=O)NC1=C(I)C(C(O)=O)=C(I)C(NC(C)=O)=C1I

Specifications

Color White
Quantity 25 g
Merck Index 14,2992
Solubility Information Soluble in Methanol.
Formula Weight 613.9
Physical Form Powder
Chemical Name or Material Diatrizoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.