missing translation for 'onlineSavingsMsg'
Learn More

Dibenzenesulfonamide, 95%

Product Code. 15417058
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417058 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15417058 Supplier Thermo Scientific Alfa Aesar Supplier No. H60569.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2618-96-4 | C12H11NO4S2 | 297.343 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2618-96-4
Melting Point 150°C
Molecular Formula C12H11NO4S2
MDL Number MFCD00025019
Quantity 250 mg
Synonym dibenzenesulfonimide, benzenesulfonamide, n-phenylsulfonyl, dibenzenesulfonamide, bis phenylsulfonyl amine, n-phenylsulfonyl benzenesulfonamide, dibenzolsulfimid, dibenzolsulfimide, diphenylsulfonimide, benzenesulfonic acid imide, n-benzenesulfonyl benzenesulfonamide
InChI Key OVQABVAKPIYHIG-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C2=CC=CC=C2
IUPAC Name N-(benzenesulfonyl)benzenesulfonamide
Molecular Weight (g/mol) 297.343
PubChem CID 75671
Formula Weight 297.35
Percent Purity 95%
Chemical Name or Material Dibenzenesulfonamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.