missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo[a,c]anthracene, TRC

Product Code. 30134499
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134499 50 mg 50mg
18022440 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134499 Supplier Toronto Research Chemicals Supplier No. TRCD41704550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 215-58-7
Molecular Formula C22 H14
Molecular Weight (g/mol) 278.35
Synonym Benzo[b]triphenylene, 1,2:3,4-Dibenzanthracene, 1,2:3,4-Dibenzoanthracene, 2,3-Benzotriphenylene, Dibenz[a,c]anthracene, Dibenzo[a,c]anthracene
IUPAC Name benzo[b]triphenylene
SMILES c1ccc2cc3c4ccccc4c5ccccc5c3cc2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H14/c1-2-8-16-14-22-20-12-6-4-10-18(20)17-9-3-5-11-19(17)21(22)13-15(16)7-1/h1-14H
Formula Weight 278.1096
Chemical Name or Material Dibenzo[a,c]anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.