missing translation for 'onlineSavingsMsg'
Learn More

Dibenzosuberenol, 96%

Product Code. 11371497
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11371497 25 g 25g
11361497 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371497 Supplier Thermo Scientific Alfa Aesar Supplier No. L02062.14
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10354-00-4 | C15H12O | 208.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10354-00-4
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00003586
InChI Key SRIISEYIFDTFRZ-UHFFFAOYSA-N
Synonym dibenzosuberenol, 5h-dibenzo a,d 7 annulen-5-ol, 5h-dibenzo a,d cyclohepten-5-ol, unii-s9t01jbh4a, s9t01jbh4a, 5h-dibenzo a,d-1-cyclohepten-5-ol, dibenzo b,f cyclohepten-1-ol, 5h-dibenzo-a,d-cyclohepten-5-ol, 5-hydrodibenzo a,d 7 annulen-5-ol
PubChem CID 82577
IUPAC Name 11H-dibenzo[1,2-a
SMILES C1=CC=C2C(C3=CC=CC=C3C=CC2=C1)O

Specifications

Melting Point 125°C to 127°C
Quantity 25 g
Beilstein 1957458
Formula Weight 208.26
Percent Purity 96%
Chemical Name or Material Dibenzosuberenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.