Learn More
Dibenzosuberone, 97%
Descrizione
Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.
Solubility
Soluble in water (0.03g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifica
Especificaciones
| Melting Point | 32°C to 34°C |
| Density | 1.156 |
| Boiling Point | 147°C to 149°C (0.3 mmHg) |
| Flash Point | 200°C (392°F) |
| Refractive Index | 1.634 |
| Quantity | 25 g |
| Beilstein | 1454355 |
| Solubility Information | Soluble in water (0.03g/L at 20°C). |
| IUPAC Name | 5,6-dihydrodibenzo[2,1-b |
| Formula Weight | 208.26 |
| Mostrar más |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.