missing translation for 'onlineSavingsMsg'
Learn More

Dibenzosuberone, TRC

Product Code. 30133356
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133356 100 mg 100mg
30003787 5 g 5g
30003961 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133356 Supplier Toronto Research Chemicals Supplier No. TRCD416940100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1210-35-1
Molecular Formula C15 H12 O
Molecular Weight (g/mol) 208.26
Synonym 10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-one, Cyproheptadine Hydrochloride Imp. B (EP), Nortriptyline Hydrochloride Imp. A (EP), Amitriptyline Hydrochloride Imp. A (EP), Dibenzosuberone
IUPAC Name 5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulen-11-one
SMILES O=C1c2ccccc2CCc3ccccc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H12O/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-8H,9-10H2
Formula Weight 208.0888
Percent Purity >95%
Chemical Name or Material Dibenzosuberone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.