missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dibenzoyl-L-tartaric acid monohydrate, 98%

Product Code. 10790851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10790851 100 g 100g
10438340 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10790851 Supplier Thermo Scientific Acros Supplier No. 160251000
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62708-56-9 | C18H14O8 | 358.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62708-56-9 , 2743-38-6
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00149119
InChI Key YONLFQNRGZXBBF-OKILXGFUSA-N
Synonym 2r,3r-2,3-bis benzoyloxy succinic acid hydrate, --dibenzoyl-l-tartaric acid monohydrate, --dibenzoyl-l-tartaric acidmonohydrate, dibenzoyl-l---tartaric acid monohydrate, l--dibenzoyl-l-tartatic acid monohydrate, 2r,3r---dibenzoyl-l-tartaric acid monohydrate, 2r,3r-2,3-bis benzoyloxy butanedioic acid hydrate, dibenzoyl-l-tartaric acid monohydrate, l-dbta, pubchem7152
PubChem CID 44119738
SMILES OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 88°C to 90°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula [C6H5CO2CH(CO2H)-]2·H2O
Quantity 100 g
Specific Rotation Condition −115° (20°C c=5,ethanol)
Specific Rotation -115
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ether and alcohol,practically insoluble in chloroform
IUPAC Name (2R,3S)-2,3-dibenzoyloxybutanedioic acid;hydrate
Formula Weight 376.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (-)-Dibenzoyl-L-tartaric acid monohydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (-)-Dibenzoyl-L-tartaric acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.