missing translation for 'onlineSavingsMsg'
Learn More

Dibenzoyl peroxide, 75%, remainder water

Product Code. 10236460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10236460 5 g Plastic bottle 5g
10702641 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10236460 Supplier Thermo Scientific Acros Supplier No. 211780050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 94-36-0 | C14H10O4 | 242.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-36-0 , 7732-18-5
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
MDL Number MFCD00003071
InChI Key OMPJBNCRMGITSC-UHFFFAOYSA-N
Synonym benzoyl peroxide, dibenzoyl peroxide, peroxide, dibenzoyl, benzoperoxide, benzoyl superoxide, acetoxyl, lucidol, benoxyl, panoxyl, benzol peroxide
PubChem CID 7187
ChEBI CHEBI:82405
IUPAC Name benzoyl benzenecarboperoxoate
SMILES C1=CC=C(C=C1)C(=O)OOC(=O)C2=CC=CC=C2

Specifications

Melting Point 104.0°C to 106.0°C
Color White
Packaging Plastic bottle
Linear Formula (C6H5CO)2O2
Quantity 5 g
Beilstein 09, 179
Fieser 01,196; 04,122; 05,182; 06,160; 07,89; 12,157; 15,146; 17,367
Merck Index 15, 1119
Solubility Information Solubility in water: insoluble. Other solubilities: sparingly soluble in alcohol,soluble in benzene,chloroform,ether and acetone,1g/40 mL carbon disulfide,1g/40 mL olive oil
Formula Weight 242.23
Percent Purity 75%
Physical Form Wet Granular Powder
Chemical Name or Material Dibenzoyl peroxide, 75%, remainder water
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dibenzoyl peroxide, 75%, remainder water
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Organic peroxide Type C & D
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H242-Heating may cause a fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.