missing translation for 'onlineSavingsMsg'
Learn More

Dibenzoyl Thiamine, MedChemExpress

Product Code. 16428876
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428876 Supplier Medchem Express Supplier No. HYB2212

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Dibenzoyl Thiamine (Bentiamine), a derivative of thiamine, is rapidly absorbed into the body and converted to thiamine.

Dibenzoyl Thiamine (Bentiamine), a derivative of thiamine, is rapidly absorbed into the body and converted to thiamine.

Specifications

Chemical Name or Material Dibenzoyl Thiamine
CAS 299-88-7
Color White
Purity Grade Notes Research
Molecular Formula C26H26N4O4S
Quantity 100 mg
Synonym Bentiamine
Solubility Information DMSO : 50 mg/mL (101.92 mM; Need ultrasonic)
SMILES O=C(C1=CC=CC=C1)S/C(CCOC(C2=CC=CC=C2)=O)=C(N(CC3=CN=C(C)N=C3N)C=O)/C
Molecular Weight (g/mol) 490.57
Formula Weight 490.57
Percent Purity 98.97%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.