missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl ketoxime, 98+%

Product Code. 11334364
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334364 25 g 25g
11324364 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334364 Supplier Thermo Scientific Alfa Aesar Supplier No. L00914.14
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1788-31-4 | C15H15NO | 225.291 g/mol

Dibenzyl ketoxime, is used as an intermediate for the preparation of cis-2-benzyl-3-phenylaziridine by reaction with lithium aluminum hydride. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl ketoxime, is used as an intermediate for the preparation of cis-2-benzyl-3-phenylaziridine by reaction with lithium aluminum hydride. It is also used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1788-31-4
Molecular Formula C15H15NO
Molecular Weight (g/mol) 225.291
MDL Number MFCD00015453
InChI Key SXEBLVKLMOIGER-UHFFFAOYSA-N
Synonym dibenzyl ketoxime, 1,3-diphenylacetone oxime, 1,3-diphenylpropan-2-one oxime, 2-propanone, 1,3-diphenyl-, oxime, n-1,3-diphenylpropan-2-ylidene hydroxylamine, 2-hydroxyimino-1,3-diphenylpropane, dibenzylketoxim, maybridge4_000477, dibenzyl ketoxime, 98+%, 1,3-diphenylacetone oxime #
PubChem CID 74518
IUPAC Name N-(1,3-diphenylpropan-2-ylidene)hydroxylamine
SMILES C1=CC=C(C=C1)CC(=NO)CC2=CC=CC=C2

Specifications

Melting Point 122°C to 124°C
Quantity 25 g
Beilstein 1965977
Solubility Information Insoluble in water.
Formula Weight 225.29
Percent Purity ≥98%
Chemical Name or Material Dibenzyl ketoxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.