missing translation for 'onlineSavingsMsg'
Learn More

Dibenzylamine, TRC

Product Code. 30359179
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359179 1 g 1g
30352883 100 g 100g
30356039 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359179 Supplier Toronto Research Chemicals Supplier No. TRCD4175051G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-49-1
Molecular Formula C14 H15 N
Molecular Weight (g/mol) 197.28
Synonym Dibenzylamine (8CI), N-(Phenylmethyl)benzenemethanamine, (N-Benzylaminomethyl)benzene, Bibenzylamine, Bisbenzylamine, DBA, N,N-Dibenzylamine, N-Benzylbenzylamine, NSC 4811
IUPAC Name N-benzyl-1-phenylmethanamine
SMILES C(NCc1ccccc1)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H15N/c1-3-7-13(8-4-1)11-15-12-14-9-5-2-6-10-14/h1-10,15H,11-12H2
Formula Weight 197.1204
Chemical Name or Material Dibenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.