Learn More
Dicer Antibody (CL0378), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP2-30699
 View more versions of this product
Product Code. 18730134
  • 518.00 € / 0.10mL
Description
Dicer Monoclonal specifically detects Dicer in Human, Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Purified
IgG2a