missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer

Product Code. 15447005
Click to view available options
Quantity:
500 mg
2 g
Unit Size:
2g
500mg
This item is not returnable. View return policy

Product Code. 15447005

Brand: Thermo Scientific Alfa Aesar 045565.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 12354-84-6 | C20H30Cl4Ir2 | 796.694 g/mol

Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer is used as a precursor to catalysts for the asymmetric transfer hydrogenation of ketones. It is a catalyst for greener amine synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12354-84-6
Molecular Formula C20H30Cl4Ir2
Molecular Weight (g/mol) 796.694
MDL Number MFCD00075435
InChI Key MMAGMBCAIFVRGJ-UHFFFAOYSA-J
Synonym unii-6oy4uuc534, dichloro pentamethylcyclopentadienyl iridium iii dimer, pentamethylcyclopentadienyl iridium iii dichloride dimer, pentamethylcyclopentadienyliridium dichloride, pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer, eta5-pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer mi, bis mu-chloro chloro eta5-pentamethylcyclopentadienyl iridium, iridium, di-mu-chlorodichlorobis 1,2,3,4,5-eta-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl di
PubChem CID 76030743
IUPAC Name iridium(3+);1,2,3,4,5-pentamethylcyclopenta-1,3-diene;tetrachloride
SMILES C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3]

Specifications

Quantity 2 g
Formula Weight 796.67
Chemical Name or Material Dichloro (pentamethylcyclopentadienyl)iridium (III) dimer
compliance-icons
Product Identifier
  • Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.