missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodiphenylmethane, 97%

Product Code. p-105356 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
Product Code. Quantity Packaging unitSize
10752695 10 mL Glass bottle 10mL
10417490 50 mL Glass bottle 50mL
10335890 250 mL Glass bottle 250mL
3 options
This item is not returnable. View return policy

Product Code. 10752695

Brand: Thermo Scientific Acros 113330100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2051-90-3 | C13H10Cl2 | 237.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-90-3
Molecular Formula C13H10Cl2
Molecular Weight (g/mol) 237.13
MDL Number MFCD00000811
InChI Key OPTDDWCXQQYKGU-UHFFFAOYSA-N
Synonym dichlorodiphenylmethane, diphenyldichloromethane, dpm halocarbon, benzophenone dichloride, alpha,alpha-dichlorodiphenylmethane, benzene, 1,1'-dichloromethylene bis, methane, dichlorodiphenyl, dichlorodiphenyl methane, dichloro phenyl methyl benzene, unii-81fez29ak6
PubChem CID 16327
IUPAC Name [dichloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(Cl)Cl

Specifications

Color Yellow
Density 1.2300g/mL
Boiling Point 305.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2CCl2
Refractive Index 1.6030 to 1.6050
Quantity 10 mL
Beilstein 05, 590
Specific Gravity 1.23
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ether,benzene,ethanol and toluene
Formula Weight 237.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dichlorodiphenylmethane, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichlorodiphenylmethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.